missing translation for 'onlineSavingsMsg'
Learn More

Methyl beta-D-Glucopyranoside Hemihydrate 98.0+%, TCI America™

Catalog No. M07095G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M07095G 5 g
M070925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M07095G Supplier TCI America Supplier No. M07095G

Chemical Identifiers

CAS 709-50-2
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00006602
InChI Key HOVAGTYPODGVJG-UHFFFAOYNA-N
Synonym Methyl beta-D-Glucoside
PubChem CID 445238
ChEBI CHEBI:320055
IUPAC Name 2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
SMILES COC1OC(CO)C(O)C(O)C1O

Specifications

Melting Point 111°C
Color White
Quantity 5 g
Formula Weight 194.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl beta-D-Glucopyranoside Hemihydrate