missing translation for 'onlineSavingsMsg'
Learn More

Methyl beta-D-galactopyranoside

Catalog No. AAL0812706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0812706 5 g
AAL0812703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0812706 Supplier Thermo Scientific Chemicals Supplier No. L0812706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1824-94-8 | C7H14O6 | 194.18 g/mol

It is a weak substrate and effective inducer of beta-D-galactosidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a weak substrate and effective inducer of beta-D-galactosidase.

Solubility
Soluble in water and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Keep it refrigerated.

Chemical Identifiers

CAS 1824-94-8
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00064357
InChI Key HOVAGTYPODGVJG-VOQCIKJUSA-N
Synonym methyl beta-d-galactopyranoside, methyl beta-d-galactoside, methyl galactoside, methylgalactoside, methyl beta-galactoside, galbetaome, unii-64ryd088rj, beta-d-galactopyranose methyl glycoside, beta-methyl-d-galactoside, methyl-beta-galactose
PubChem CID 94214
ChEBI CHEBI:17540
IUPAC Name (2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
SMILES CO[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 175°C to 179°C
Quantity 5 g
Beilstein 81569
Solubility Information Soluble in water and methanol.
Optical Rotation −16° (c=1.5 in Methanol)
Formula Weight 194.18
Chemical Name or Material Methyl beta-D-galactopyranoside
EINECSNumber 217-361-7
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only