missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl beta-D-Galactopyranoside 98.0+%, TCI America™
$89.48 - $244.19
Chemical Identifiers
| CAS | 1824-94-8 |
|---|---|
| Molecular Formula | C7H14O6 |
| Molecular Weight (g/mol) | 194.18 |
| MDL Number | MFCD00064357 |
| InChI Key | HOVAGTYPODGVJG-VOQCIKJUSA-N |
| Synonym | methyl beta-d-galactopyranoside, methyl beta-d-galactoside, methyl galactoside, methylgalactoside, methyl beta-galactoside, galbetaome, unii-64ryd088rj, beta-d-galactopyranose methyl glycoside, beta-methyl-d-galactoside, methyl-beta-galactose |
| PubChem CID | 94214 |
| ChEBI | CHEBI:17540 |
| IUPAC Name | (2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol |
| SMILES | CO[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O |
Chemical Identifiers
| 1824-94-8 | |
| 194.18 | |
| HOVAGTYPODGVJG-VOQCIKJUSA-N | |
| 94214 | |
| (2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol |
| C7H14O6 | |
| MFCD00064357 | |
| methyl beta-d-galactopyranoside, methyl beta-d-galactoside, methyl galactoside, methylgalactoside, methyl beta-galactoside, galbetaome, unii-64ryd088rj, beta-d-galactopyranose methyl glycoside, beta-methyl-d-galactoside, methyl-beta-galactose | |
| CHEBI:17540 | |
| CO[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O |
Specifications
| 1824-94-8 | |
| White | |
| MFCD00064357 | |
| methyl beta-d-galactopyranoside, methyl beta-d-galactoside, methyl galactoside, methylgalactoside, methyl beta-galactoside, galbetaome, unii-64ryd088rj, beta-d-galactopyranose methyl glycoside, beta-methyl-d-galactoside, methyl-beta-galactose | |
| CO[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O | |
| 194.18 | |
| CHEBI:17540 | |
| ≥98.0% (GC) | |
| Methyl beta-D-Galactopyranoside |
| 178°C | |
| C7H14O6 | |
| 1 g | |
| HOVAGTYPODGVJG-VOQCIKJUSA-N | |
| (2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol | |
| 94214 | |
| 194.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No