missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl beta-D-Galactopyranoside 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 178°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 194.18 |
| Percent Purity | ≥98.0% (GC) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Methyl beta-D-Galactopyranoside |
Safety and Handling
| TSCA | No |