missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl-beta-cyclodextrin (mixture of several Methylated), TCI America™
Supplier: TCI America M1356250G
| Quantity | 250 g |
|---|
Chemical Identifiers
| 128446-36-6 | |
| 1303.311 | |
| YZOUYRAONFXZSI-SBHWVFSVSA-N | |
| 51051622 |
| C54H94O35 | |
| MFCD00074980 | |
| beta-cyclodextrin methyl ethers, beta-cyd, methyl-?cyclodextrin, beta-w7 m1.8, dimethyl-, a-cyclodextrin, .beta.-cyclodextrin,methylethers, methyl-b-cyclodextrin cell culture*tested | |
| COC1C(C2C(OC1OC3C(OC(C(C3OC)OC)OC4C(OC(C(C4OC)OC)OC5C(OC(C(C5OC)OC)OC6C(OC(C(C6OC)OC)OC7C(OC(C(C7OC)OC)OC8C(OC(O2)C(C8O)OC)CO)CO)CO)CO)CO)CO)CO)O |
Specifications
| Methyl-beta-cyclodextrin (mixture of several Methylated) | |
| C54H94O35 | |
| MFCD00074980 | |
| YZOUYRAONFXZSI-SBHWVFSVSA-N | |
| 1303.311 | |
| Crystalline Powder |
| 128446-36-6 | |
| 250 g | |
| beta-cyclodextrin methyl ethers, beta-cyd, methyl-?cyclodextrin, beta-w7 m1.8, dimethyl-, a-cyclodextrin, .beta.-cyclodextrin,methylethers, methyl-b-cyclodextrin cell culture*tested | |
| COC1C(C2C(OC1OC3C(OC(C(C3OC)OC)OC4C(OC(C(C4OC)OC)OC5C(OC(C(C5OC)OC)OC6C(OC(C(C6OC)OC)OC7C(OC(C(C7OC)OC)OC8C(OC(O2)C(C8O)OC)CO)CO)CO)CO)CO)CO)CO)O | |
| 51051622 |
Safety and Handling
TSCA : Yes