missing translation for 'onlineSavingsMsg'
Learn More

Methyl Benzenesulfonate 98.0+%, TCI America™

Catalog No. B003325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B003325ML 25 mL
B0033100ML 100 mL
B0033500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B003325ML Supplier TCI America Supplier No. B003325ML

Chemical Identifiers

CAS 80-18-2
Molecular Formula C7H8O3S
Molecular Weight (g/mol) 172.20
MDL Number MFCD00014737
InChI Key CZXGXYBOQYQXQD-UHFFFAOYSA-N
Synonym benzenesulfonic acid, methyl ester, methyl benzenesulphonate, methyl besylate, methylbenzenesulfonate, ccris 9156, benzenesulfonic acid methyl ester, methyl benzene sulfonate, methyl ester of benzenesulphonic acid, methylbenzene sulfonate, pubchem22730
PubChem CID 6630
IUPAC Name methyl benzenesulfonate
SMILES COS(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point -4°C
Color Yellow
Boiling Point 280°C
Quantity 25 mL
Formula Weight 172.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Benzenesulfonate
RTECSNumber DB7151000
TSCA Yes