missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Azetidine-3-carboxylate Hydrochloride 98.0+%, TCI America™
$57.79 - $165.90
Chemical Identifiers
| CAS | 100202-39-9 |
|---|---|
| Molecular Formula | C5H10ClNO2 |
| Molecular Weight (g/mol) | 151.59 |
| MDL Number | MFCD01861758 |
| InChI Key | UOCWTLBPYROHEF-UHFFFAOYSA-N |
| Synonym | methyl azetidine-3-carboxylate hydrochloride, azetidine-3-methyl carboxylate hydrochloride, azetidine-3-carboxylic acid methyl ester hydrochloride, 3-azetidinecarboxylic acid, methyl ester, hydrochloride, methyl azetidine-3-carboxylate hcl, methylazetidine-3-carboxylate hydrochloride, methyl 3-azetidinecarboxylate hydrochloride, azetidine-3-carboxylic acid methyl ester hcl, pubchem23516, acmc-209tka |
| PubChem CID | 21100040 |
| IUPAC Name | methyl azetidine-3-carboxylate hydrochloride |
| SMILES | Cl.COC(=O)C1CNC1 |
Chemical Identifiers
| 100202-39-9 | |
| 151.59 | |
| UOCWTLBPYROHEF-UHFFFAOYSA-N | |
| 21100040 | |
| Cl.COC(=O)C1CNC1 |
| C5H10ClNO2 | |
| MFCD01861758 | |
| methyl azetidine-3-carboxylate hydrochloride, azetidine-3-methyl carboxylate hydrochloride, azetidine-3-carboxylic acid methyl ester hydrochloride, 3-azetidinecarboxylic acid, methyl ester, hydrochloride, methyl azetidine-3-carboxylate hcl, methylazetidine-3-carboxylate hydrochloride, methyl 3-azetidinecarboxylate hydrochloride, azetidine-3-carboxylic acid methyl ester hcl, pubchem23516, acmc-209tka | |
| methyl azetidine-3-carboxylate hydrochloride |
Specifications
| 100202-39-9 | |
| White | |
| C5H10ClNO2 | |
| 1 g | |
| UOCWTLBPYROHEF-UHFFFAOYSA-N | |
| methyl azetidine-3-carboxylate hydrochloride | |
| 21100040 | |
| ≥98.0% (T) | |
| Methyl Azetidine-3-carboxylate Hydrochloride |
| 95°C | |
| 192°C | |
| MFCD01861758 | |
| methyl azetidine-3-carboxylate hydrochloride, azetidine-3-methyl carboxylate hydrochloride, azetidine-3-carboxylic acid methyl ester hydrochloride, 3-azetidinecarboxylic acid, methyl ester, hydrochloride, methyl azetidine-3-carboxylate hcl, methylazetidine-3-carboxylate hydrochloride, methyl 3-azetidinecarboxylate hydrochloride, azetidine-3-carboxylic acid methyl ester hcl, pubchem23516, acmc-209tka | |
| Cl.COC(=O)C1CNC1 | |
| 151.59 | |
| 151.59 | |
| Crystalline Powder |
Safety and Handling
TSCA : No