missing translation for 'onlineSavingsMsg'
Learn More

Methyl Azetidine-3-carboxylate Hydrochloride 98.0+%, TCI America™

Catalog No. M25211G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25211G 1 g
M25215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25211G Supplier TCI America Supplier No. M25211G

Chemical Identifiers

CAS 100202-39-9
Molecular Formula C5H10ClNO2
Molecular Weight (g/mol) 151.59
MDL Number MFCD01861758
InChI Key UOCWTLBPYROHEF-UHFFFAOYSA-N
Synonym methyl azetidine-3-carboxylate hydrochloride, azetidine-3-methyl carboxylate hydrochloride, azetidine-3-carboxylic acid methyl ester hydrochloride, 3-azetidinecarboxylic acid, methyl ester, hydrochloride, methyl azetidine-3-carboxylate hcl, methylazetidine-3-carboxylate hydrochloride, methyl 3-azetidinecarboxylate hydrochloride, azetidine-3-carboxylic acid methyl ester hcl, pubchem23516, acmc-209tka
PubChem CID 21100040
IUPAC Name methyl azetidine-3-carboxylate hydrochloride
SMILES Cl.COC(=O)C1CNC1

Specifications

Melting Point 95°C
Color White
Boiling Point 192°C
Quantity 1 g
Formula Weight 151.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Azetidine-3-carboxylate Hydrochloride