Learn More
Methyl azetidine-3-carboxylate hydrochloride, 95%, Thermo Scientific Chemicals
$280.68 - $280.68
Chemical Identifiers
| CAS | 100202-39-9 |
|---|---|
| Molecular Formula | C5H10ClNO2 |
| Molecular Weight (g/mol) | 151.59 |
| MDL Number | MFCD01861758 |
| InChI Key | UOCWTLBPYROHEF-UHFFFAOYSA-N |
| Synonym | methyl azetidine-3-carboxylate hydrochloride, azetidine-3-methyl carboxylate hydrochloride, azetidine-3-carboxylic acid methyl ester hydrochloride, 3-azetidinecarboxylic acid, methyl ester, hydrochloride, methyl azetidine-3-carboxylate hcl, methylazetidine-3-carboxylate hydrochloride, methyl 3-azetidinecarboxylate hydrochloride, azetidine-3-carboxylic acid methyl ester hcl, pubchem23516, acmc-209tka |
| PubChem CID | 21100040 |
| IUPAC Name | methyl azetidine-3-carboxylate;hydrochloride |
| SMILES | Cl.COC(=O)C1CNC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH57489ME
|
Thermo Scientific Chemicals
H57489ME |
500 mg |
Each for $280.68
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 100202-39-9 | |
| 151.59 | |
| UOCWTLBPYROHEF-UHFFFAOYSA-N | |
| 21100040 | |
| Cl.COC(=O)C1CNC1 |
| C5H10ClNO2 | |
| MFCD01861758 | |
| methyl azetidine-3-carboxylate hydrochloride, azetidine-3-methyl carboxylate hydrochloride, azetidine-3-carboxylic acid methyl ester hydrochloride, 3-azetidinecarboxylic acid, methyl ester, hydrochloride, methyl azetidine-3-carboxylate hcl, methylazetidine-3-carboxylate hydrochloride, methyl 3-azetidinecarboxylate hydrochloride, azetidine-3-carboxylic acid methyl ester hcl, pubchem23516, acmc-209tka | |
| methyl azetidine-3-carboxylate;hydrochloride |
Specifications
| 100202-39-9 | |
| 69°C (156°F) | |
| MFCD01861758 | |
| methyl azetidine-3-carboxylate hydrochloride, azetidine-3-methyl carboxylate hydrochloride, azetidine-3-carboxylic acid methyl ester hydrochloride, 3-azetidinecarboxylic acid, methyl ester, hydrochloride, methyl azetidine-3-carboxylate hcl, methylazetidine-3-carboxylate hydrochloride, methyl 3-azetidinecarboxylate hydrochloride, azetidine-3-carboxylic acid methyl ester hcl, pubchem23516, acmc-209tka | |
| Cl.COC(=O)C1CNC1 | |
| 151.59 | |
| 151.59 | |
| Methyl azetidine-3-carboxylate hydrochloride |
| 187°C to 192°C | |
| C5H10ClNO2 | |
| 500 mg | |
| UOCWTLBPYROHEF-UHFFFAOYSA-N | |
| methyl azetidine-3-carboxylate;hydrochloride | |
| 21100040 | |
| 95% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only