missing translation for 'onlineSavingsMsg'
Learn More

Methyl azetidine-3-carboxylate hydrochloride, 95%, Thermo Scientific Chemicals

Catalog No. AAH57489ME
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57489ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH57489ME Supplier Thermo Scientific Chemicals Supplier No. H57489ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 100202-39-9 | C5H10ClNO2 | 151.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100202-39-9
Molecular Formula C5H10ClNO2
Molecular Weight (g/mol) 151.59
MDL Number MFCD01861758
InChI Key UOCWTLBPYROHEF-UHFFFAOYSA-N
Synonym methyl azetidine-3-carboxylate hydrochloride, azetidine-3-methyl carboxylate hydrochloride, azetidine-3-carboxylic acid methyl ester hydrochloride, 3-azetidinecarboxylic acid, methyl ester, hydrochloride, methyl azetidine-3-carboxylate hcl, methylazetidine-3-carboxylate hydrochloride, methyl 3-azetidinecarboxylate hydrochloride, azetidine-3-carboxylic acid methyl ester hcl, pubchem23516, acmc-209tka
PubChem CID 21100040
IUPAC Name methyl azetidine-3-carboxylate;hydrochloride
SMILES Cl.COC(=O)C1CNC1

Specifications

Boiling Point 187°C to 192°C
Flash Point 69°C (156°F)
Quantity 500 mg
Formula Weight 151.59
Percent Purity 95%
Chemical Name or Material Methyl azetidine-3-carboxylate hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only