missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Angelate 97.0+%, TCI America™
Supplier: TCI America A112625ML
Specifications
| Methyl Angelate | |
| Yellow | |
| C6H10O2 | |
| 25 mL | |
| Angelic Acid Methyl Ester, (Z)-2-Methyl-2-butenoic Acid Methyl Ester, 2-Methylisocrotonic Acid Methyl Ester, Methyl (Z)-2-Methyl-2-butenoate, Methyl 2-Methylisocrotonate | |
| CC=C(C)C(=O)OC | |
| 114.144 | |
| 114.14 | |
| Liquid |
| 5953-76-4 | |
| 130°C | |
| MFCD00063645 | |
| 3272 | |
| YYJWBYNQJLBIGS-PLNGDYQASA-N | |
| methyl (Z)-2-methylbut-2-enoate | |
| 5356352 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 5953-76-4 | |
| 114.144 | |
| YYJWBYNQJLBIGS-PLNGDYQASA-N | |
| 5356352 | |
| CC=C(C)C(=O)OC |
| C6H10O2 | |
| MFCD00063645 | |
| Angelic Acid Methyl Ester, (Z)-2-Methyl-2-butenoic Acid Methyl Ester, 2-Methylisocrotonic Acid Methyl Ester, Methyl (Z)-2-Methyl-2-butenoate, Methyl 2-Methylisocrotonate | |
| methyl (Z)-2-methylbut-2-enoate |
Safety and Handling
EINECSNumber : (2)-3326
TSCA : No
Recommended Storage : Refrigerator