missing translation for 'onlineSavingsMsg'
Learn More

Methyl alpha-L-rhamnopyranoside, 98%

Catalog No. p-7027405
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2412906 5 g
AAB2412914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2412906 Supplier Thermo Scientific Chemicals Supplier No. B2412906

CAS: 14917-55-6 | C7H14O5 | 178.184 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 14917-55-6
Molecular Formula C7H14O5
Molecular Weight (g/mol) 178.184
MDL Number MFCD00067654
InChI Key OHWCAVRRXKJCRB-PAMBMQIZSA-N
Synonym methyl alpha-l-rhamnopyranoside, methyl-alpha-l-rhamnopyranoside, 1-o-methyl-alpha-rhamnose, methyl 6-deoxy-alpha-l-mannopyranoside, methyl a-l-rhamnopyranoside, rao, methyl rhamnopyranoside, 2r,3r,4r,5r,6s-2-methoxy-6-methyloxane-3,4,5-triol, 2r,3r,4r,5r,6s-2-methoxy-6-methyltetrahydro-2h-pyran-3,4,5-triol, methyl-a-l-rhamnopyranoside
PubChem CID 84695
IUPAC Name (2R,3R,4R,5R,6S)-2-methoxy-6-methyloxane-3,4,5-triol
SMILES CC1C(C(C(C(O1)OC)O)O)O

Specifications

Melting Point 106°C to 108°C
Quantity 5 g
Formula Weight 178.19
Percent Purity 98%
Chemical Name or Material Methyl alpha-L-rhamnopyranoside
EINECSNumber 238-987-7
TSCA No

RUO – Research Use Only