missing translation for 'onlineSavingsMsg'
Learn More

Methyl alpha-D-xylopyranoside, 98%

Catalog No. p-7043233
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6640603 1 g
AAJ6640606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6640603 Supplier Thermo Scientific Chemicals Supplier No. J6640603

CAS: 91-09-8 | C6H12O5 | 164.157 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 91-09-8
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.157
MDL Number MFCD00053593
InChI Key ZBDGHWFPLXXWRD-MOJAZDJTSA-N
Synonym methyl alpha-d-xylopyranoside, alpha-d-xylopyranoside, methyl, methyl a-d-xylopyranoside, 2s,3r,4s,5r-2-methoxyoxane-3,4,5-triol, 1-o-methyl-alpha-d-xylopyranoside, beta-methyl-d-xyloside, methyl beta-d-xylopyranoside, methyl-alpha-d-xylopyranoside, alpha methyl-d-xyloside, methyl, a-d-xylopyranoside
PubChem CID 101554
IUPAC Name (2S,3R,4S,5R)-2-methoxyoxane-3,4,5-triol
SMILES COC1C(C(C(CO1)O)O)O

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in water
Optical Rotation +154° (c=10 in Water)
Formula Weight 264.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Methyl alpha-D-xylopyranoside
EINECSNumber 202-040-6
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only