missing translation for 'onlineSavingsMsg'
Learn More

Methyl-alpha-D-mannopyranoside, 99%

CAS: 617-04-9 | C7H14O6 | 194.18 g/mol

$97.33 - $275.07

Chemical Identifiers

CAS 617-04-9
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00063262
InChI Key HOVAGTYPODGVJG-UHFFFAOYNA-N
Synonym methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
PubChem CID 101798
ChEBI CHEBI:43943
IUPAC Name (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
SMILES COC1OC(CO)C(O)C(O)C1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1153314
View Documents
Thermo Scientific Chemicals
A1153314
25 g
Each for $97.33
Only null left
 
AAA1153322
View Documents
Thermo Scientific Chemicals
A1153322
100 g
Each for $275.07
Only null left
 
Description

Description

Methyl a-D-mannopyranoside Is a competitor inhibitor of the binding of mannose by Escherichia coli. It is also used as a pharmaceutical raw material, health care product and intermediate. It is also been used as Animal Pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl a-D-mannopyranoside Is a competitor inhibitor of the binding of mannose by Escherichia coli. It is also used as a pharmaceutical raw material, health care product and intermediate. It is also been used as Animal Pharmaceuticals.

Solubility
It is easily soluble in methanol, water. Insoluble in ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agent. Stable under recommended storage conditions
Specifications

Chemical Identifiers

617-04-9
194.18
HOVAGTYPODGVJG-UHFFFAOYNA-N
101798
(2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
C7H14O6
MFCD00063262
methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
CHEBI:43943
COC1OC(CO)C(O)C(O)C1O

Specifications

617-04-9
C7H14O6
25 g
methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
HOVAGTYPODGVJG-UHFFFAOYNA-N
COC1OC(CO)C(O)C(O)C1O
194.18
CHEBI:43943
99%
193°C to 196°C
MFCD00063262
81566
It is easily soluble in methanol,water. Insoluble in ether.
+79° (c=10 in water)
(2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
101798
194.18
Methyl-alpha-D-mannopyranoside
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 210-502-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only