missing translation for 'onlineSavingsMsg'
Learn More

Methyl alpha-D-glucopyranoside, 98%

Catalog No. p-7020502
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1248422 100 g
AAA1248436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1248422 Supplier Thermo Scientific Chemicals Supplier No. A1248422

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 97-30-3 | C7H14O6 | 194.183 g/mol

Methyl alpha-D-glucopyranoside is a biochemical used to vary echo decay times in a study that assessed the effects of cryoprotection on the structure and activity of p21ras. It is also used in a study to investigate saccharide-mediated protection of chaotropic-induced deactivation of concanavalin A.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl alpha-D-glucopyranoside is a biochemical used to vary echo decay times in a study that assessed the effects of cryoprotection on the structure and activity of p21ras. It is also used in a study to investigate saccharide-mediated protection of chaotropic-induced deactivation of concanavalin A.

Solubility
Soluble in water. Insoluble in ether

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature. Store under dry inert gas.

Chemical Identifiers

CAS 97-30-3
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.183
MDL Number MFCD00064086
InChI Key HOVAGTYPODGVJG-IECVIRLLSA-N
Synonym methyl alpha-d-glucopyranoside, methyl alpha-d-glucoside, alpha-methylglucoside, alpha-methyl-d-glucoside, alpha-methyl d-glucose ether, methyl-alpha-d-glucopyranoside, alpha-d-methylglucoside, me alpha-glc, alpha-d-methyl glucoside, unii-qcf122nf3r
PubChem CID 7568545
IUPAC Name (2R,3R,4R,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
SMILES COC1C(C(C(C(O1)CO)O)O)O

Specifications

Melting Point 166°C to 171°C
Density 1.46
Quantity 100 g
Beilstein 81568
Sensitivity Hygroscopic
Merck Index 14,6080
Solubility Information Soluble in water; Insoluble in ether
Optical Rotation +159° (c=10 in Water)
Formula Weight 194.18
Percent Purity ≥98%
Chemical Name or Material Methyl alpha-D-glucopyranoside
Show More Show Less
EINECSNumber 202-571-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only