missing translation for 'onlineSavingsMsg'
Learn More

Methyl alpha-chloroacrylate, 98+%, stabilized with hydroquinone

Catalog No. AC414160250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC414160250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC414160250 Supplier Thermo Scientific Chemicals Supplier No. 414160250

CAS: 80-63-7 | C4H5ClO2 | 120.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80-63-7
Molecular Formula C4H5ClO2
Molecular Weight (g/mol) 120.54
MDL Number MFCD00051368
InChI Key AWJZTPWDQYFQPQ-UHFFFAOYSA-N
Synonym methyl 2-chloroacrylate, methyl alpha-chloroacrylate, methyl-2-chloroacrylate, 2-propenoic acid, 2-chloro-, methyl ester, methyl-alpha-chloroacrylate, 2-chloroacrylic acid, methyl ester, methyl 2-chloro-2-propenoate, acrylic acid, 2-chloro-, methyl ester, methyl 2-chloro-2-propenate, unii-47pg4l077j
PubChem CID 6659
IUPAC Name methyl 2-chloroprop-2-enoate
SMILES COC(=O)C(=C)Cl

Specifications

Color Colorless to Brown
Density 1.1890g/mL
Boiling Point 52°C (50.0 mmHg)
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4420 to 1.444
Linear Formula CH2CClCOOCH3
Quantity 25 g
Specific Gravity 1.189
Solubility Information Solubility in water: .. Other solubilities: soluble in diethylether
Stabilizer abt. 0.06% hydroquinone
Formula Weight 120.54
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Methyl α-chloroacrylate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 201-298-7

RUO – Research Use Only