missing translation for 'onlineSavingsMsg'
Learn More

Methyl Alcohol, Exceeds A.C.S Specifications, HPLC Grade, 99.9%, Spectrum™ Chemical

Catalog No. p-7157585
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 4 L
6 x 1 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604409 1 L Amber Glass Bottle
18604410 4 L Amber Glass Bottle
18604411 6 x 1 L Amber Glass Bottle
18604412 4 x 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604409 Supplier Spectrum Chemical Mfg Cor Supplier No. HP7021LT
Only null left

HP702, 67-56-1, CH3OH

Spectrum™ Chemical Methyl Alcohol, Exceeds A.C.S Specifications, HPLC Grade is a colorless, flammable substance used in chemical labs as a polar protic solvent in organic synthesis, as well as a carrier (mobile phase) solvent in high pressure liquid chromatography (HPLC). Less expensive than Acetonitrile, the popularity Methyl Alcohol HPLC continues to grow, especially in reversed phase HPLC.

All HPLC and Ultra-pure solvents offered by Spectrum are glass distilled, submicron filtered, and undergo rigorous specification testing to insure accurate, repeatable results.

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
IUPAC Name methanol
SMILES CO

Specifications

CAS Max % 100%
Quantity 1 L
Packaging Amber Glass Bottle
Residue after Evaporation 2 ppm
Titratable Acid 0.0003 meq/g
Titratable Base 0.0002 meq/g
UN Number UN1230
Water 0.05%
Boiling Point 64°C to 65°C
Percent Purity 99.9%
Grade High Purity
Chemical Name or Material Methyl Alcohol
Show More Show Less