missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 9-Hydroxynonanoate 95.0+%, TCI America™
$480.92 - $1433.46
Chemical Identifiers
| CAS | 34957-73-8 |
|---|---|
| Molecular Formula | C10H20O3 |
| Molecular Weight (g/mol) | 188.267 |
| MDL Number | MFCD00191518 |
| InChI Key | RIZOOQYPYGPBOC-UHFFFAOYSA-N |
| Synonym | 9-Hydroxypelargonic Acid Methyl Ester, 9-Hydroxynonanoic Acid Methyl Ester, Methyl 9-Hydroxypelargonate |
| PubChem CID | 292983 |
| IUPAC Name | methyl 9-hydroxynonanoate |
| SMILES | COC(=O)CCCCCCCCO |
Chemical Identifiers
| 34957-73-8 | |
| 188.267 | |
| RIZOOQYPYGPBOC-UHFFFAOYSA-N | |
| 292983 | |
| COC(=O)CCCCCCCCO |
| C10H20O3 | |
| MFCD00191518 | |
| 9-Hydroxypelargonic Acid Methyl Ester, 9-Hydroxynonanoic Acid Methyl Ester, Methyl 9-Hydroxypelargonate | |
| methyl 9-hydroxynonanoate |
Specifications
| 34957-73-8 | |
| 272°C | |
| MFCD00191518 | |
| 9-Hydroxypelargonic Acid Methyl Ester, 9-Hydroxynonanoic Acid Methyl Ester, Methyl 9-Hydroxypelargonate | |
| COC(=O)CCCCCCCCO | |
| 188.267 | |
| 188.27 | |
| Liquid |
| Yellow | |
| C10H20O3 | |
| 1 g | |
| RIZOOQYPYGPBOC-UHFFFAOYSA-N | |
| methyl 9-hydroxynonanoate | |
| 292983 | |
| ≥95.0% (GC) | |
| Methyl 9-Hydroxynonanoate |
Safety and Handling
TSCA : No