missing translation for 'onlineSavingsMsg'
Learn More

Methyl 9-Hydroxynonanoate 95.0+%, TCI America™

Catalog No. H08351G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08351G 1 g
H08355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08351G Supplier TCI America Supplier No. H08351G
Only null left

Chemical Identifiers

CAS 34957-73-8
Molecular Formula C10H20O3
Molecular Weight (g/mol) 188.267
MDL Number MFCD00191518
InChI Key RIZOOQYPYGPBOC-UHFFFAOYSA-N
Synonym 9-Hydroxypelargonic Acid Methyl Ester, 9-Hydroxynonanoic Acid Methyl Ester, Methyl 9-Hydroxypelargonate
PubChem CID 292983
IUPAC Name methyl 9-hydroxynonanoate
SMILES COC(=O)CCCCCCCCO

Specifications

Color Yellow
Boiling Point 272°C
Quantity 1 g
Formula Weight 188.27
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 9-Hydroxynonanoate