missing translation for 'onlineSavingsMsg'
Learn More

Methyl 9-Formylnonanoate 95.0+%, TCI America™

Catalog No. M22631G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22631G 1 g
M22635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22631G Supplier TCI America Supplier No. M22631G

Chemical Identifiers

CAS 14811-73-5
Molecular Formula C11H20O3
Molecular Weight (g/mol) 200.278
InChI Key LVPNLDGNIRVDRF-UHFFFAOYSA-N
Synonym 9-Formylnonanoic Acid Methyl Ester
PubChem CID 519005
IUPAC Name methyl 10-oxodecanoate
SMILES COC(=O)CCCCCCCCC=O

Specifications

Color Yellow
Boiling Point 121°C
Quantity 1 g
Formula Weight 200.28
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 9-Formylnonanoate
TSCA No
Recommended Storage Refrigerator