missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-methoxyindole-2-carboxylate, 97%

Catalog No. AAH6652806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6652806 5 g
AAH6652803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6652806 Supplier Thermo Scientific Chemicals Supplier No. H6652806

CAS: 98081-83-5 | C11H11NO3 | 205.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98081-83-5
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.21
MDL Number MFCD00134301
InChI Key OPUUCOLVBDQWEY-UHFFFAOYSA-N
Synonym methyl 6-methoxy-2-indolecarboxylate, 6-methoxy-1h-indole-2-carboxylic acid methyl ester, methyl-6-methoxy-2-indolecarboxylate, methyl 6-methoxy-indole-2-carboxylate, 6-methoxyindole-2-carboxylic acid methyl ester, methyl 6-methoxyindole-2-carboxylate, 1h-indole-2-carboxylic acid, 6-methoxy-, methyl ester, methyl 6-methoxy-2-indolecarboxylat, pubchem7251, 6-methoxy-2-indolecarboxylate
PubChem CID 688173
IUPAC Name methyl 6-methoxy-1H-indole-2-carboxylate
SMILES COC(=O)C1=CC2=CC=C(OC)C=C2N1

Specifications

Melting Point 117°C to 119°C
Quantity 5 g
Beilstein 395862
Formula Weight 205.21
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 6-methoxyindole-2-carboxylate
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only