missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-Hydroxynicotinate 97.0+%, TCI America™

Catalog No. p-7133392
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27271G 1 g
M27275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27271G Supplier TCI America Supplier No. M27271G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66171-50-4
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD08436011
InChI Key JTVVPKLHKMKWNN-UHFFFAOYSA-N
Synonym 6-Hydroxynicotinic Acid Methyl Ester, Methyl 6-Hydroxy-3-pyridinecarboxylate, 6-Hydroxy-3-pyridinecarboxylic Acid Methyl Ester
PubChem CID 416343
IUPAC Name methyl 6-oxo-1H-pyridine-3-carboxylate
SMILES COC(=O)C1=CNC(=O)C=C1

Specifications

Melting Point 166°C
Color White-Yellow
Quantity 1 g
Formula Weight 153.14
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 6-Hydroxynicotinate