missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-Fluoronicotinate 98.0+%, TCI America™

Catalog No. M27245G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27245G 5 g
M27241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27245G Supplier TCI America Supplier No. M27245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1427-06-1
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD03095098
InChI Key HLYBWNNPVXFCPZ-UHFFFAOYSA-N
Synonym methyl 6-fluoronicotinate, 6-fluoronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 6-fluoro-, methyl ester, 6-fluoronicotinicacidmethylester, methyl6-fluoropyridine-3-carboxylate, 6-fluoro-3-methoxycarbonyl pyridine, 6-fluoro-nicotinic acid methyl ester, 6-fluoro-3-pyridinecarboxylic acid methyl ester, pubchem10629, methyl6-fluoronicotinate
PubChem CID 13758765
IUPAC Name methyl 6-fluoropyridine-3-carboxylate
SMILES COC(=O)C1=CN=C(C=C1)F

Specifications

Melting Point 51°C
Color White-Yellow
Quantity 5 g
Formula Weight 155.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 6-Fluoronicotinate