missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-fluoronicotinate, 95%, Thermo Scientific Chemicals

Catalog No. AAH6130203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6130203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6130203 Supplier Thermo Scientific Chemicals Supplier No. H6130203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1427-06-1 | Molecular Formula: C7H6FNO2 | Molecular Weight: 155.128 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1427-06-1
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD03095098
InChI Key HLYBWNNPVXFCPZ-UHFFFAOYSA-N
PubChem CID 13758765
IUPAC Name methyl 6-fluoropyridine-3-carboxylate
SMILES COC(=O)C1=CN=C(C=C1)F

Specifications

Melting Point 48°C to 52°C
Flash Point 99°C (210°F)
Quantity 1 g
Synonym methyl 6-fluoronicotinate, 6-fluoronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 6-fluoro-, methyl ester, 6-fluoronicotinicacidmethylester, methyl6-fluoropyridine-3-carboxylate, 6-fluoro-3-methoxycarbonyl pyridine, 6-fluoro-nicotinic acid methyl ester, 6-fluoro-3-pyridinecarboxylic acid methyl ester, pubchem10629, methyl6-fluoronicotinate
Formula Weight 155.13
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl 6-fluoronicotinate
Hazard Category H302-H318
Hazard Statement GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only