missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-chloropyridine-2-carboxylate, 95%

Catalog No. AAH6402314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6402314 25 g
AAH6402306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6402314 Supplier Thermo Scientific Chemicals Supplier No. H6402314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6636-55-1 | C7H6ClNO2 | 171.58 g/mol

Methyl 6-chloropyridine-2-carboxylate was coupled with 2-(trimethylstannyl)pyridine to afford methyl 2,2?-bipyridine-6-carboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 6-chloropyridine-2-carboxylate was coupled with 2-(trimethylstannyl)pyridine to afford methyl 2,2′-bipyridine-6-carboxylate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.

Chemical Identifiers

CAS 6636-55-1
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00155391
InChI Key TWUXBVMXSBEKHA-UHFFFAOYSA-N
Synonym methyl 6-chloropicolinate, methyl 6-chloro-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 6-chloro-, methyl ester, 2-chloropyridine-6-carboxylate methyl, 6-chloro-2-picolinic acid methyl ester, 6-chloropicolinic acid methyl ester, methyl 2-chloro-6-pyridinecarboxylate, methyl 2-chloropyridine-6-carboxylate, 6-chloro-pyridine-2-carboxylic acid methyl ester, 6-chloro-2-pyridinecarboxylic acid methyl ester
PubChem CID 242723
IUPAC Name methyl 6-chloropyridine-2-carboxylate
SMILES COC(=O)C1=NC(=CC=C1)Cl

Specifications

Melting Point 92°C to 98°C (literature)
Color Yellow
Quantity 25 g
Beilstein 128580
Solubility Information Slightly soluble in water.
Formula Weight 171.58
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl 6-chloropyridine-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only