missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-Chloro-2-pyridinecarboxylate 98.0+%, TCI America™

Catalog No. M26405G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26405G 5 g
M26401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26405G Supplier TCI America Supplier No. M26405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6636-55-1 | C7H6ClNO2 | 171.58 g/mol

Chemical Identifiers

CAS 6636-55-1
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00155391
InChI Key TWUXBVMXSBEKHA-UHFFFAOYSA-N
Synonym methyl 6-chloropicolinate, methyl 6-chloro-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 6-chloro-, methyl ester, 2-chloropyridine-6-carboxylate methyl, 6-chloro-2-picolinic acid methyl ester, 6-chloropicolinic acid methyl ester, methyl 2-chloro-6-pyridinecarboxylate, methyl 2-chloropyridine-6-carboxylate, 6-chloro-pyridine-2-carboxylic acid methyl ester, 6-chloro-2-pyridinecarboxylic acid methyl ester
PubChem CID 242723
IUPAC Name methyl 6-chloropyridine-2-carboxylate
SMILES COC(=O)C1=NC(=CC=C1)Cl

Specifications

Melting Point 95°C
Color White
Quantity 5 g
Formula Weight 171.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 6-Chloro-2-pyridinecarboxylate