missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 6-Aminopyridine-2-carboxylate 98.0+%, TCI America™
$48.13 - $188.67
Chemical Identifiers
| CAS | 36052-26-3 |
|---|---|
| Molecular Formula | C7H8N2O2 |
| Molecular Weight (g/mol) | 152.153 |
| MDL Number | MFCD00233712 |
| InChI Key | OHIHEJTUXNQOPM-UHFFFAOYSA-N |
| Synonym | 6-Aminopyridine-2-carboxylic Acid Methyl Ester, Methyl 6-Aminopicolinate, 6-Aminopicolinic Acid Methyl Ester |
| PubChem CID | 13809143 |
| IUPAC Name | methyl 6-aminopyridine-2-carboxylate |
| SMILES | COC(=O)C1=NC(=CC=C1)N |
Chemical Identifiers
| 36052-26-3 | |
| 152.153 | |
| OHIHEJTUXNQOPM-UHFFFAOYSA-N | |
| 13809143 | |
| COC(=O)C1=NC(=CC=C1)N |
| C7H8N2O2 | |
| MFCD00233712 | |
| 6-Aminopyridine-2-carboxylic Acid Methyl Ester, Methyl 6-Aminopicolinate, 6-Aminopicolinic Acid Methyl Ester | |
| methyl 6-aminopyridine-2-carboxylate |
Specifications
| 36052-26-3 | |
| White-Yellow | |
| MFCD00233712 | |
| 6-Aminopyridine-2-carboxylic Acid Methyl Ester, Methyl 6-Aminopicolinate, 6-Aminopicolinic Acid Methyl Ester | |
| COC(=O)C1=NC(=CC=C1)N | |
| 152.153 | |
| 152.15 | |
| Crystalline Powder |
| 97°C | |
| C7H8N2O2 | |
| 1 g | |
| OHIHEJTUXNQOPM-UHFFFAOYSA-N | |
| methyl 6-aminopyridine-2-carboxylate | |
| 13809143 | |
| ≥98.0% (GC) | |
| Methyl 6-Aminopyridine-2-carboxylate |
Safety and Handling
TSCA : No