missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 6-Aminopyridine-2-carboxylate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 97°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 152.15 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Methyl 6-Aminopyridine-2-carboxylate |
Safety and Handling
| TSCA | No |