missing translation for 'onlineSavingsMsg'
Learn More

Methyl-5-norbornene-2,3-dicarboxylic Anhydride (mixture of isomers) 80.0+%, TCI America™

Catalog No. M069025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M069025G 25 g
M0690500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M069025G Supplier TCI America Supplier No. M069025G
Only null left

Chemical Identifiers

CAS 25134-21-8
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00167036
InChI Key KNRCVAANTQNTPT-UHFFFAOYNA-N
Synonym Methyl-3,6-endomethylenetetrahydrophthalic Anhydride, Methylnadic Anhydride

Specifications

Color Yellow
Quantity 25 g
UN Number 3265
Formula Weight 178.19
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl-5-norbornene-2,3-dicarboxylic Anhydride (mixture of isomers)
EINECSNumber (4)-0298
RTECSNumber RB9100000
TSCA Yes