missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 5-Methylnicotinate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 48°C |
| Color | White-Yellow |
| Boiling Point | 90°C |
| Quantity | 5 g |
| Synonym | methyl 5-methylnicotinate, methyl5-methylnicotinate, 5-methylnicotinic acid methyl ester, 5-methyl-nicotinic acid methyl ester, methyl 5-methyl nicotinate, 3-pyridinecarboxylic acid, 5-methyl-, methyl ester, 3-methoxycarbonyl-5-methylpyridine, 5-methylpyridine-3-carboxylic acid methyl ester, 5-methylnicotin methyl ester, acmc-1clej |
| Formula Weight | 151.17 |
| Percent Purity | ≥98.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Methyl 5-Methylnicotinate |
Safety and Handling
| TSCA | No |