missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-Methylnicotinate 98.0+%, TCI America™

Catalog No. M24281G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24281G 1 g
M24285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24281G Supplier TCI America Supplier No. M24281G

Chemical Identifiers

CAS 29681-45-6
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD01075739
InChI Key KQILMMLAGGFMCM-UHFFFAOYSA-N
PubChem CID 256308
IUPAC Name methyl 5-methylpyridine-3-carboxylate
SMILES CC1=CN=CC(=C1)C(=O)OC

Specifications

Melting Point 48°C
Color White-Yellow
Boiling Point 90°C
Quantity 1 g
Synonym methyl 5-methylnicotinate, methyl5-methylnicotinate, 5-methylnicotinic acid methyl ester, 5-methyl-nicotinic acid methyl ester, methyl 5-methyl nicotinate, 3-pyridinecarboxylic acid, 5-methyl-, methyl ester, 3-methoxycarbonyl-5-methylpyridine, 5-methylpyridine-3-carboxylic acid methyl ester, 5-methylnicotin methyl ester, acmc-1clej
Formula Weight 151.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-Methylnicotinate