missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-methylnicotinate, 94%

CAS: 29681-45-6 | C8H9NO2 | 151.165 g/mol

Supplier:  Thermo Scientific Chemicals L1637106

 View more versions of this product

Catalog No. AAL1637106


Only null left
Explore available promotions

Description

Description

5-Methyl-N?-nitrosonornicotine (5-MeNNN) was synthesized from methyl 5-methylnicotinate. 5-Methyl-N?-nitrosonornicotine is used as the starting material in organic compound synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-N′-nitrosonornicotine (5-MeNNN) was synthesized from methyl 5-methylnicotinate. 5-Methyl-N′-nitrosonornicotine is used as the starting material in organic compound synthesis.

Solubility
Soluble in Chloroform and Methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

Methyl 5-methylnicotinate
42°C to 46°C
C8H9NO2
5 g
methyl 5-methylnicotinate, methyl5-methylnicotinate, 5-methylnicotinic acid methyl ester, 5-methyl-nicotinic acid methyl ester, methyl 5-methyl nicotinate, 3-pyridinecarboxylic acid, 5-methyl-, methyl ester, 3-methoxycarbonyl-5-methylpyridine, 5-methylpyridine-3-carboxylic acid methyl ester, 5-methylnicotin methyl ester, acmc-1clej
KQILMMLAGGFMCM-UHFFFAOYSA-N
methyl 5-methylpyridine-3-carboxylate
256308
94%
29681-45-6
90°C (3 mmHg)
MFCD01075739
472498
Soluble in Chloroform and Methanol.
CC1=CN=CC(=C1)C(=O)OC
151.165
151.17

Chemical Identifiers

29681-45-6
151.165
KQILMMLAGGFMCM-UHFFFAOYSA-N
methyl 5-methylpyridine-3-carboxylate
C8H9NO2
MFCD01075739
256308
CC1=CN=CC(=C1)C(=O)OC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only