missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-methylnicotinate, 94%

Catalog No. AAL1637103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1637103 1 g
AAL1637106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1637103 Supplier Thermo Scientific Chemicals Supplier No. L1637103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 29681-45-6 | C8H9NO2 | 151.165 g/mol

5-Methyl-N?-nitrosonornicotine (5-MeNNN) was synthesized from methyl 5-methylnicotinate. 5-Methyl-N?-nitrosonornicotine is used as the starting material in organic compound synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-N′-nitrosonornicotine (5-MeNNN) was synthesized from methyl 5-methylnicotinate. 5-Methyl-N′-nitrosonornicotine is used as the starting material in organic compound synthesis.

Solubility
Soluble in Chloroform and Methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 29681-45-6
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD01075739
InChI Key KQILMMLAGGFMCM-UHFFFAOYSA-N
PubChem CID 256308
IUPAC Name methyl 5-methylpyridine-3-carboxylate
SMILES CC1=CN=CC(=C1)C(=O)OC

Specifications

Melting Point 42°C to 46°C
Boiling Point 90°C (3 mmHg)
Quantity 1 g
Beilstein 472498
Synonym methyl 5-methylnicotinate, methyl5-methylnicotinate, 5-methylnicotinic acid methyl ester, 5-methyl-nicotinic acid methyl ester, methyl 5-methyl nicotinate, 3-pyridinecarboxylic acid, 5-methyl-, methyl ester, 3-methoxycarbonyl-5-methylpyridine, 5-methylpyridine-3-carboxylic acid methyl ester, 5-methylnicotin methyl ester, acmc-1clej
Solubility Information Soluble in Chloroform and Methanol.
Formula Weight 151.17
Percent Purity 94%
Chemical Name or Material Methyl 5-methylnicotinate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only