missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-methylisoxazole-3-carboxylate, 97%

Catalog No. AAA1981414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1981414 25 g
AAA1981422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1981414 Supplier Thermo Scientific Chemicals Supplier No. A1981414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 19788-35-3 | C6H7NO3 | 141.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19788-35-3
Molecular Formula C6H7NO3
Molecular Weight (g/mol) 141.13
MDL Number MFCD00015895
InChI Key MVHHQOCEOUNTID-UHFFFAOYSA-N
Synonym methyl 5-methylisoxazole-3-carboxylate, methyl 5-methyl-3-isoxazolecarboxylate, 3-isoxazolecarboxylic acid, 5-methyl-, methyl ester, 5-methyl-3-isoxazolecarboxylic acid methyl ester, methyl 5-methyl-3-isoxazole carboxylate, methyl-5-methyl-3-isoxazole carboxylate, 5-methylisoxazole-3-carboxylic acid methyl ester, 5-methyl-isoxazole-3-carboxylic acid methyl ester, acmc-209xf5, ksc489s1r
PubChem CID 88245
IUPAC Name methyl 5-methyl-1,2-oxazole-3-carboxylate
SMILES COC(=O)C1=NOC(C)=C1

Specifications

Melting Point 91°C to 92°C
Boiling Point 125°C (11 mmHg)
Quantity 25 g
Beilstein 115704
Formula Weight 141.13
Percent Purity 97%
Chemical Name or Material Methyl 5-methylisoxazole-3-carboxylate
Hazard Category H500
EINECSNumber 243-311-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only