missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-(chloromethyl)-2-furoate, 95%, Thermo Scientific™

$243.11 - $243.11

Chemical Identifiers

CAS 2144-37-8
Molecular Formula C7H7ClO3
Molecular Weight (g/mol) 174.58
MDL Number MFCD00020922
InChI Key PWXMEBZOKUPCST-UHFFFAOYSA-N
Synonym methyl 5-chloromethyl-2-furoate, methyl 5-chloromethyl furan-2-carboxylate, methyl 5-chloromethyl-2-furancarboxylate, 5-chloromethyl-furan-2-carboxylic acid methyl ester, 2-furancarboxylic acid, 5-chloromethyl-, methyl ester, 5-chloromethylfuran-2-carboxylic acid methyl ester, pubchem24261, enamine_005466, acmc-1ck5d
PubChem CID 75065
IUPAC Name methyl 5-(chloromethyl)furan-2-carboxylate
SMILES COC(=O)C1=CC=C(O1)CCl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BLT00257EA
SDS View Documents
Thermo Scientific Chemicals
BLT00257EA
10 g
Each for $243.11
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2144-37-8
174.58
PWXMEBZOKUPCST-UHFFFAOYSA-N
75065
COC(=O)C1=CC=C(O1)CCl
C7H7ClO3
MFCD00020922
methyl 5-chloromethyl-2-furoate, methyl 5-chloromethyl furan-2-carboxylate, methyl 5-chloromethyl-2-furancarboxylate, 5-chloromethyl-furan-2-carboxylic acid methyl ester, 2-furancarboxylic acid, 5-chloromethyl-, methyl ester, 5-chloromethylfuran-2-carboxylic acid methyl ester, pubchem24261, enamine_005466, acmc-1ck5d
methyl 5-(chloromethyl)furan-2-carboxylate

Specifications

2144-37-8
100.0
Amber Glass Bottle
MFCD00020922
methyl 5-chloromethyl-2-furoate, methyl 5-chloromethyl furan-2-carboxylate, methyl 5-chloromethyl-2-furancarboxylate, 5-chloromethyl-furan-2-carboxylic acid methyl ester, 2-furancarboxylic acid, 5-chloromethyl-, methyl ester, 5-chloromethylfuran-2-carboxylic acid methyl ester, pubchem24261, enamine_005466, acmc-1ck5d
COC(=O)C1=CC=C(O1)CCl
174.58
174.58
Methyl 5-(chloromethyl)-2-furoate
94.5
95%
C7H7ClO3
10 g
PWXMEBZOKUPCST-UHFFFAOYSA-N
methyl 5-(chloromethyl)furan-2-carboxylate
75065
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'