Learn More
Methyl 5-bromopyrimidine-2-carboxylate, 95%, Thermo Scientific Chemicals
$88.79 - $268.33
Chemical Identifiers
| CAS | 89581-38-4 |
|---|---|
| Molecular Formula | C6H5BrN2O2 |
| Molecular Weight (g/mol) | 217.02 |
| MDL Number | MFCD11111606 |
| InChI Key | XILAKTMDKMVJQV-UHFFFAOYSA-N |
| Synonym | methyl 5-bromopyrimidine-2-carboxyate, methyl-5-bromo-2 pyrimidine carboxylate, 5-bromo-pyrimidine-2-carboxylic acid methyl ester, 2-pyrimidinecarboxylic acid, 5-bromo-, methyl ester, 2-pyrimidinecarboxylicacid, 5-bromo-, methyl ester, 5-bromopyrimidine-2-carboxylic acid methyl ester, acmc-20dmbr, pubchem21032, methyl5-bromopyrimidine-2-carboxylate, methyl 5-bromanylpyrimidine-2-carboxylate |
| PubChem CID | 45790831 |
| IUPAC Name | methyl 5-bromopyrimidine-2-carboxylate |
| SMILES | COC(=O)C1=NC=C(Br)C=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH61339MD
|
Thermo Scientific Chemicals
H61339MD |
250 mg |
Each for $88.79
|
|
|||||
|
AAH6133903
|
Thermo Scientific Chemicals
H6133903 |
1 g |
Each for $268.33
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 89581-38-4 | |
| 217.02 | |
| XILAKTMDKMVJQV-UHFFFAOYSA-N | |
| 45790831 | |
| COC(=O)C1=NC=C(Br)C=N1 |
| C6H5BrN2O2 | |
| MFCD11111606 | |
| methyl 5-bromopyrimidine-2-carboxyate, methyl-5-bromo-2 pyrimidine carboxylate, 5-bromo-pyrimidine-2-carboxylic acid methyl ester, 2-pyrimidinecarboxylic acid, 5-bromo-, methyl ester, 2-pyrimidinecarboxylicacid, 5-bromo-, methyl ester, 5-bromopyrimidine-2-carboxylic acid methyl ester, acmc-20dmbr, pubchem21032, methyl5-bromopyrimidine-2-carboxylate, methyl 5-bromanylpyrimidine-2-carboxylate | |
| methyl 5-bromopyrimidine-2-carboxylate |
Specifications
| 89581-38-4 | |
| C6H5BrN2O2 | |
| 250 mg | |
| XILAKTMDKMVJQV-UHFFFAOYSA-N | |
| methyl 5-bromopyrimidine-2-carboxylate | |
| 45790831 | |
| 95% | |
| Methyl 5-bromopyrimidine-2-carboxylate |
| White | |
| MFCD11111606 | |
| methyl 5-bromopyrimidine-2-carboxyate, methyl-5-bromo-2 pyrimidine carboxylate, 5-bromo-pyrimidine-2-carboxylic acid methyl ester, 2-pyrimidinecarboxylic acid, 5-bromo-, methyl ester, 2-pyrimidinecarboxylicacid, 5-bromo-, methyl ester, 5-bromopyrimidine-2-carboxylic acid methyl ester, acmc-20dmbr, pubchem21032, methyl5-bromopyrimidine-2-carboxylate, methyl 5-bromanylpyrimidine-2-carboxylate | |
| COC(=O)C1=NC=C(Br)C=N1 | |
| 217.02 | |
| 217.02 | |
| Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only