missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 5-Bromopyridine-2-carboxylate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 100°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Synonym | methyl 5-bromopicolinate, 5-bromopyridine-2-carboxylic acid methyl ester, methyl 5-bromo-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 5-bromo-, methyl ester, 5-bromo-2-pyridinecarboxylic acid methyl ester, 5-bromo-pyridine-2-carboxylic acid methyl ester, methyl5-bromopicolinate, pubchem10589, 5-bromopyridine-2-carboxylicacidmethylester, acmc-1cjr7 |
| Formula Weight | 216.03 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Methyl 5-Bromopyridine-2-carboxylate |
Safety and Handling
| TSCA | No |