missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-Bromopyridine-2-carboxylate 98.0+%, TCI America™

Catalog No. M26141G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26141G 1 g
M26145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26141G Supplier TCI America Supplier No. M26141G
Only null left

Chemical Identifiers

CAS 29682-15-3
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD04112493
InChI Key JEURNBCYNWNADN-UHFFFAOYSA-N
PubChem CID 7016458
IUPAC Name methyl 5-bromopyridine-2-carboxylate
SMILES COC(=O)C1=NC=C(C=C1)Br

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 1 g
Synonym methyl 5-bromopicolinate, 5-bromopyridine-2-carboxylic acid methyl ester, methyl 5-bromo-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 5-bromo-, methyl ester, 5-bromo-2-pyridinecarboxylic acid methyl ester, 5-bromo-pyridine-2-carboxylic acid methyl ester, methyl5-bromopicolinate, pubchem10589, 5-bromopyridine-2-carboxylicacidmethylester, acmc-1cjr7
Formula Weight 216.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-Bromopyridine-2-carboxylate