missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 5-Bromonicotinate, TCI America™

$41.25 - $125.28

Chemical Identifiers

CAS 29681-44-5
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00173668
InChI Key AAJZXPWBILCHAW-UHFFFAOYSA-N
Synonym methyl 5-bromonicotinate, 5-bromonicotinic acid methyl ester, 5-bromo-3-pyridinecarboxylic acid methyl ester, 5-bromo nicotinic acid methyl ester, 3-pyridinecarboxylic acid, 5-bromo-, methyl ester, 3-bromo-5-methoxycarbonyl pyridine, 5-bromopyridine-3-carboxylic acid methyl ester, 5-bromo-nicotinic acid methyl ester, methyl 5-bromo-3-pyridinecarboxylate, methyl 3-bromo-5-pyridinecarboxylate
PubChem CID 699336
IUPAC Name methyl 5-bromopyridine-3-carboxylate
SMILES COC(=O)C1=CC(=CN=C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M20081G View Documents TCI America
M20081G
1g Each for $41.25
 
M20085G View Documents TCI America
M20085G
5g Each for $125.28
 
Specifications

Chemical Identifiers

29681-44-5
216.034
AAJZXPWBILCHAW-UHFFFAOYSA-N
699336
COC(=O)C1=CC(=CN=C1)Br
C7H6BrNO2
MFCD00173668
methyl 5-bromonicotinate, 5-bromonicotinic acid methyl ester, 5-bromo-3-pyridinecarboxylic acid methyl ester, 5-bromo nicotinic acid methyl ester, 3-pyridinecarboxylic acid, 5-bromo-, methyl ester, 3-bromo-5-methoxycarbonyl pyridine, 5-bromopyridine-3-carboxylic acid methyl ester, 5-bromo-nicotinic acid methyl ester, methyl 5-bromo-3-pyridinecarboxylate, methyl 3-bromo-5-pyridinecarboxylate
methyl 5-bromopyridine-3-carboxylate

Specifications

White-Yellow
1g
C7H6BrNO2
methyl 5-bromonicotinate, 5-bromonicotinic acid methyl ester, 5-bromo-3-pyridinecarboxylic acid methyl ester, 5-bromo nicotinic acid methyl ester, 3-pyridinecarboxylic acid, 5-bromo-, methyl ester, 3-bromo-5-methoxycarbonyl pyridine, 5-bromopyridine-3-carboxylic acid methyl ester, 5-bromo-nicotinic acid methyl ester, methyl 5-bromo-3-pyridinecarboxylate, methyl 3-bromo-5-pyridinecarboxylate
COC(=O)C1=CC(=CN=C1)Br
216.034
216.03
≥98.0% (GC,T)
99°C
29681-44-5
MFCD00173668
AAJZXPWBILCHAW-UHFFFAOYSA-N
methyl 5-bromopyridine-3-carboxylate
699336
Crystal-Powder at 20°C
Methyl 5-Bromonicotinate
Safety and Handling

Safety and Handling

TSCA : No