Learn More
Methyl 5-bromonicotinate, 97%, Thermo Scientific Chemicals
$108.21 - $108.21
Chemical Identifiers
| CAS | 29681-44-5 |
|---|---|
| Molecular Formula | C7H6BrNO2 |
| Molecular Weight (g/mol) | 216.034 |
| MDL Number | MFCD00173668 |
| InChI Key | AAJZXPWBILCHAW-UHFFFAOYSA-N |
| Synonym | methyl 5-bromonicotinate, 5-bromonicotinic acid methyl ester, 5-bromo-3-pyridinecarboxylic acid methyl ester, 5-bromo nicotinic acid methyl ester, 3-pyridinecarboxylic acid, 5-bromo-, methyl ester, 3-bromo-5-methoxycarbonyl pyridine, 5-bromopyridine-3-carboxylic acid methyl ester, 5-bromo-nicotinic acid methyl ester, methyl 5-bromo-3-pyridinecarboxylate, methyl 3-bromo-5-pyridinecarboxylate |
| PubChem CID | 699336 |
| IUPAC Name | methyl 5-bromopyridine-3-carboxylate |
| SMILES | COC(=O)C1=CC(=CN=C1)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2090806
|
Thermo Scientific Chemicals
B2090806 |
5 g |
Each for $108.21
|
|
|||||
Description
A substrate used in a microwave-assisted, palladium-catalyzed arylation of acetone as its TMS enol ether.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA substrate used in a microwave-assisted, palladium-catalyzed arylation of acetone as its TMS enol ether.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 29681-44-5 | |
| 216.034 | |
| AAJZXPWBILCHAW-UHFFFAOYSA-N | |
| 699336 | |
| COC(=O)C1=CC(=CN=C1)Br |
| C7H6BrNO2 | |
| MFCD00173668 | |
| methyl 5-bromonicotinate, 5-bromonicotinic acid methyl ester, 5-bromo-3-pyridinecarboxylic acid methyl ester, 5-bromo nicotinic acid methyl ester, 3-pyridinecarboxylic acid, 5-bromo-, methyl ester, 3-bromo-5-methoxycarbonyl pyridine, 5-bromopyridine-3-carboxylic acid methyl ester, 5-bromo-nicotinic acid methyl ester, methyl 5-bromo-3-pyridinecarboxylate, methyl 3-bromo-5-pyridinecarboxylate | |
| methyl 5-bromopyridine-3-carboxylate |
Specifications
| 29681-44-5 | |
| C7H6BrNO2 | |
| 5 g | |
| Slightly soluble in water. | |
| COC(=O)C1=CC(=CN=C1)Br | |
| 216.034 | |
| 216.04 | |
| Methyl 5-bromonicotinate |
| 96°C to 98°C | |
| MFCD00173668 | |
| methyl 5-bromonicotinate, 5-bromonicotinic acid methyl ester, 5-bromo-3-pyridinecarboxylic acid methyl ester, 5-bromo nicotinic acid methyl ester, 3-pyridinecarboxylic acid, 5-bromo-, methyl ester, 3-bromo-5-methoxycarbonyl pyridine, 5-bromopyridine-3-carboxylic acid methyl ester, 5-bromo-nicotinic acid methyl ester, methyl 5-bromo-3-pyridinecarboxylate, methyl 3-bromo-5-pyridinecarboxylate | |
| AAJZXPWBILCHAW-UHFFFAOYSA-N | |
| methyl 5-bromopyridine-3-carboxylate | |
| 699336 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
RUO – Research Use Only