missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromonicotinate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2090806

 View more versions of this product

Catalog No. AAB2090806

Only null left
Explore available promotions

Description

Description

A substrate used in a microwave-assisted, palladium-catalyzed arylation of acetone as its TMS enol ether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A substrate used in a microwave-assisted, palladium-catalyzed arylation of acetone as its TMS enol ether.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

Methyl 5-bromonicotinate
96°C to 98°C
MFCD00173668
methyl 5-bromonicotinate, 5-bromonicotinic acid methyl ester, 5-bromo-3-pyridinecarboxylic acid methyl ester, 5-bromo nicotinic acid methyl ester, 3-pyridinecarboxylic acid, 5-bromo-, methyl ester, 3-bromo-5-methoxycarbonyl pyridine, 5-bromopyridine-3-carboxylic acid methyl ester, 5-bromo-nicotinic acid methyl ester, methyl 5-bromo-3-pyridinecarboxylate, methyl 3-bromo-5-pyridinecarboxylate
AAJZXPWBILCHAW-UHFFFAOYSA-N
methyl 5-bromopyridine-3-carboxylate
699336
97%
29681-44-5
C7H6BrNO2
5 g
Slightly soluble in water.
COC(=O)C1=CC(=CN=C1)Br
216.034
216.04

Chemical Identifiers

29681-44-5
216.034
AAJZXPWBILCHAW-UHFFFAOYSA-N
699336
COC(=O)C1=CC(=CN=C1)Br
C7H6BrNO2
MFCD00173668
methyl 5-bromonicotinate, 5-bromonicotinic acid methyl ester, 5-bromo-3-pyridinecarboxylic acid methyl ester, 5-bromo nicotinic acid methyl ester, 3-pyridinecarboxylic acid, 5-bromo-, methyl ester, 3-bromo-5-methoxycarbonyl pyridine, 5-bromopyridine-3-carboxylic acid methyl ester, 5-bromo-nicotinic acid methyl ester, methyl 5-bromo-3-pyridinecarboxylate, methyl 3-bromo-5-pyridinecarboxylate
methyl 5-bromopyridine-3-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only