Learn More
Methyl 5-bromo-2-(methylthio)pyrimidine-4-carboxylate, 97%, Thermo Scientific Chemicals
$579.01 - $579.01
Chemical Identifiers
| CAS | 50593-91-4 |
|---|---|
| Molecular Formula | C7H7BrN2O2S |
| Molecular Weight (g/mol) | 263.11 |
| MDL Number | MFCD05664428 |
| InChI Key | MYZJIEWTRJTWCD-UHFFFAOYSA-N |
| Synonym | methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarboxylate, methyl 5-bromo-2-methylthio pyrimidine-4-carboxylate, methyl 5-bromo-2-methylsulfanyl pyrimidine-4-carboxylate, methyl 5-bromo-2-methylthio-4-pyrimidinecarboxylate, methylbromomethylsulfanylpyrimidinecarboxylate, methyl 5-bromo-2-methylthiopyrimidine-4-carboxylate, methyl-5-bromo-2-methylthio pyrimidine-4-carboxylate, methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarb, methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarboxylate,97, 5-bromo-2-methylsulfanylpyrimidine-4-carboxylic acid methyl ester |
| PubChem CID | 2794729 |
| SMILES | COC(=O)C1=NC(SC)=NC=C1Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6441706
|
Thermo Scientific Chemicals
H6441706 |
5 g |
Each for $579.01
|
|
|||||
Description
It can be used as pharmaceutical raw material and intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt can be used as pharmaceutical raw material and intermediate.
Solubility
Slightly soluble in water (0.42 g/L).
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
| 50593-91-4 | |
| 263.11 | |
| MYZJIEWTRJTWCD-UHFFFAOYSA-N | |
| 2794729 |
| C7H7BrN2O2S | |
| MFCD05664428 | |
| methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarboxylate, methyl 5-bromo-2-methylthio pyrimidine-4-carboxylate, methyl 5-bromo-2-methylsulfanyl pyrimidine-4-carboxylate, methyl 5-bromo-2-methylthio-4-pyrimidinecarboxylate, methylbromomethylsulfanylpyrimidinecarboxylate, methyl 5-bromo-2-methylthiopyrimidine-4-carboxylate, methyl-5-bromo-2-methylthio pyrimidine-4-carboxylate, methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarb, methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarboxylate,97, 5-bromo-2-methylsulfanylpyrimidine-4-carboxylic acid methyl ester | |
| COC(=O)C1=NC(SC)=NC=C1Br |
Specifications
| 50593-91-4 | |
| Yellow | |
| MFCD05664428 | |
| UN2811 | |
| methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarboxylate, methyl 5-bromo-2-methylthio pyrimidine-4-carboxylate, methyl 5-bromo-2-methylsulfanyl pyrimidine-4-carboxylate, methyl 5-bromo-2-methylthio-4-pyrimidinecarboxylate, methylbromomethylsulfanylpyrimidinecarboxylate, methyl 5-bromo-2-methylthiopyrimidine-4-carboxylate, methyl-5-bromo-2-methylthio pyrimidine-4-carboxylate, methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarb, methyl 5-bromo-2-methylsulfanyl-4-pyrimidinecarboxylate,97, 5-bromo-2-methylsulfanylpyrimidine-4-carboxylic acid methyl ester | |
| MYZJIEWTRJTWCD-UHFFFAOYSA-N | |
| methyl 5-bromo-2-(methylsulfanyl)pyrimidine-4-carboxylate | |
| 2794729 | |
| 97% | |
| Methyl 5-bromo-2-(methylthio)pyrimidine-4-carboxylate |
| 61°C to 64°C | |
| C7H7BrN2O2S | |
| 5 g | |
| 910703 | |
| Slightly soluble in water (0.42g/L). | |
| COC(=O)C1=NC(SC)=NC=C1Br | |
| 263.11 | |
| 263.11 | |
| Solid |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only