Learn More
Methyl 5-bromo-2-furoate, 98%, Thermo Scientific™
$271.87 - $1,148.67
Chemical Identifiers
| CAS | 2527-99-3 |
|---|---|
| Molecular Formula | C6H5BrO3 |
| Molecular Weight (g/mol) | 205.01 |
| InChI Key | FBPIDMAELBIRLE-UHFFFAOYSA-N |
| Synonym | methyl 5-bromo-2-furoate, methyl 5-bromo-2-furancarboxylate, 5-bromo-2-furoic acid methyl ester, 2-furancarboxylic acid, 5-bromo-, methyl ester, 5-bromo-2-furancarboxylic acid methyl ester, 5-bromofuran-2-carboxylic acid methyl ester, methyl 5-bromofuroate, 2-furoic acid, 5-bromo-, methyl ester, methyl 5-bromo-furanate, maybridge1_004996 |
| PubChem CID | 599633 |
| IUPAC Name | methyl 5-bromofuran-2-carboxylate |
| SMILES | COC(=O)C1=CC=C(O1)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC355750050
|
N/A | 5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC355750250
|
N/A | 25g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 2527-99-3 | |
| 205.01 | |
| methyl 5-bromo-2-furoate, methyl 5-bromo-2-furancarboxylate, 5-bromo-2-furoic acid methyl ester, 2-furancarboxylic acid, 5-bromo-, methyl ester, 5-bromo-2-furancarboxylic acid methyl ester, 5-bromofuran-2-carboxylic acid methyl ester, methyl 5-bromofuroate, 2-furoic acid, 5-bromo-, methyl ester, methyl 5-bromo-furanate, maybridge1_004996 | |
| methyl 5-bromofuran-2-carboxylate |
| C6H5BrO3 | |
| FBPIDMAELBIRLE-UHFFFAOYSA-N | |
| 599633 | |
| COC(=O)C1=CC=C(O1)Br |
Specifications
| 2527-99-3 | |
| 97.5% min. (GC) | |
| methyl 5-bromo-2-furoate, methyl 5-bromo-2-furancarboxylate, 5-bromo-2-furoic acid methyl ester, 2-furancarboxylic acid, 5-bromo-, methyl ester, 5-bromo-2-furancarboxylic acid methyl ester, 5-bromofuran-2-carboxylic acid methyl ester, methyl 5-bromofuroate, 2-furoic acid, 5-bromo-, methyl ester, methyl 5-bromo-furanate, maybridge1_004996 | |
| COC(=O)C1=CC=C(O1)Br | |
| 205.01 | |
| 205.01 | |
| Methyl 5-bromo-2-furoate |
| Authentic | |
| C6H5BrO3 | |
| FBPIDMAELBIRLE-UHFFFAOYSA-N | |
| methyl 5-bromofuran-2-carboxylate | |
| 599633 | |
| 98% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wash face,ha
GHS Signal Word: Warning
RUO – Research Use Only