missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromo-2-furoate, 97%

Catalog No. p-7025166
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2052403 1 g
AAB2052406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2052403 Supplier Thermo Scientific Chemicals Supplier No. B2052403

CAS: 2527-99-3 | C6H5BrO3 | 205.007 g/mol

It is used in the preparation of hydroquinone bis(2-carbomethoxy-5-furyl ether).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of hydroquinone bis(2-carbomethoxy-5-furyl ether).

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 2527-99-3
Molecular Formula C6H5BrO3
MDL Number MFCD00092313
InChI Key FBPIDMAELBIRLE-UHFFFAOYSA-N
Synonym methyl 5-bromo-2-furoate, methyl 5-bromo-2-furancarboxylate, 5-bromo-2-furoic acid methyl ester, 2-furancarboxylic acid, 5-bromo-, methyl ester, 5-bromo-2-furancarboxylic acid methyl ester, 5-bromofuran-2-carboxylic acid methyl ester, methyl 5-bromofuroate, 2-furoic acid, 5-bromo-, methyl ester, methyl 5-bromo-furanate, maybridge1_004996
PubChem CID 599633
IUPAC Name methyl 5-bromofuran-2-carboxylate

Specifications

Melting Point 62°C to 65°C
Quantity 1 g
Solubility Information Slightly soluble in water.
SMILES COC(=O)C1=CC=C(O1)Br
Molecular Weight (g/mol) 205.007
Formula Weight 205.01
Percent Purity 97%
Chemical Name or Material Methyl 5-bromo-2-furoate
Hazard Category H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only