missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-Bromo-2-furancarboxylate 98.0+%, TCI America™

Catalog No. M25381G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25381G 1 g
M25385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25381G Supplier TCI America Supplier No. M25381G

Chemical Identifiers

CAS 2527-99-3
Molecular Formula C6H5BrO3
Molecular Weight (g/mol) 205.007
MDL Number MFCD00092313
InChI Key FBPIDMAELBIRLE-UHFFFAOYSA-N
Synonym methyl 5-bromo-2-furoate, methyl 5-bromo-2-furancarboxylate, 5-bromo-2-furoic acid methyl ester, 2-furancarboxylic acid, 5-bromo-, methyl ester, 5-bromo-2-furancarboxylic acid methyl ester, 5-bromofuran-2-carboxylic acid methyl ester, methyl 5-bromofuroate, 2-furoic acid, 5-bromo-, methyl ester, methyl 5-bromo-furanate, maybridge1_004996
PubChem CID 599633
IUPAC Name methyl 5-bromofuran-2-carboxylate
SMILES COC(=O)C1=CC=C(O1)Br

Specifications

Melting Point 66°C
Color White-Yellow
Boiling Point 80°C
Quantity 1 g
Formula Weight 205.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-Bromo-2-furancarboxylate