missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 5-Aminolevulinate Hydrochloride 98.0+%, TCI America™
$46.20 - $202.15
Chemical Identifiers
| CAS | 79416-27-6 |
|---|---|
| Molecular Formula | C6H12ClNO3 |
| Molecular Weight (g/mol) | 181.616 |
| MDL Number | MFCD00012868 |
| InChI Key | UJYSYPVQHFNBML-UHFFFAOYSA-N |
| Synonym | methyl 5-amino-4-oxopentanoate hydrochloride, 5-aminolevulinic acid methyl ester hydrochloride, methyl aminolevulinate hydrochloride, methyl 5-aminolevulinate hydrochloride, metvixia, methyl aminolevulinate hcl, metvix pdt, 3-aminoacetylpropionic acid methyl ester hydrochloride, pentanoic acid, 5-amino-4-oxo-, methyl ester, hydrochloride, methyl aminolevulinate hydrochloride usan |
| PubChem CID | 157921 |
| ChEBI | CHEBI:60641 |
| IUPAC Name | methyl 5-amino-4-oxopentanoate;hydrochloride |
| SMILES | COC(=O)CCC(=O)CN.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A1411100MG
|
TCI America
A1411100MG |
100 mg |
Each for $46.20
|
|
|||||
|
A14111G
|
TCI America
A14111G |
1 g |
Each for $202.15
|
|
|||||
Chemical Identifiers
| 79416-27-6 | |
| 181.616 | |
| UJYSYPVQHFNBML-UHFFFAOYSA-N | |
| 157921 | |
| methyl 5-amino-4-oxopentanoate;hydrochloride |
| C6H12ClNO3 | |
| MFCD00012868 | |
| methyl 5-amino-4-oxopentanoate hydrochloride, 5-aminolevulinic acid methyl ester hydrochloride, methyl aminolevulinate hydrochloride, methyl 5-aminolevulinate hydrochloride, metvixia, methyl aminolevulinate hcl, metvix pdt, 3-aminoacetylpropionic acid methyl ester hydrochloride, pentanoic acid, 5-amino-4-oxo-, methyl ester, hydrochloride, methyl aminolevulinate hydrochloride usan | |
| CHEBI:60641 | |
| COC(=O)CCC(=O)CN.Cl |
Specifications
| 122°C | |
| White | |
| MFCD00012868 | |
| UJYSYPVQHFNBML-UHFFFAOYSA-N | |
| methyl 5-amino-4-oxopentanoate;hydrochloride | |
| 181.616 | |
| CHEBI:60641 | |
| ≥98.0% (N,T) | |
| Methyl 5-Aminolevulinate Hydrochloride |
| 79416-27-6 | |
| C6H12ClNO3 | |
| methyl 5-amino-4-oxopentanoate hydrochloride, 5-aminolevulinic acid methyl ester hydrochloride, methyl aminolevulinate hydrochloride, methyl 5-aminolevulinate hydrochloride, metvixia, methyl aminolevulinate hcl, metvix pdt, 3-aminoacetylpropionic acid methyl ester hydrochloride, pentanoic acid, 5-amino-4-oxo-, methyl ester, hydrochloride, methyl aminolevulinate hydrochloride usan | |
| COC(=O)CCC(=O)CN.Cl | |
| 100 mg | |
| 157921 | |
| 181.62 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator