missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-amino-2-furoate, 98%

Catalog No. aal0595803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0595803 1 g
AAL0595806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0595803 Supplier Thermo Scientific Chemicals Supplier No. L0595803
Only null left

CAS: 22600-30-2 | C6H7NO3 | 141.126 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22600-30-2
Molecular Formula C6H7NO3
Molecular Weight (g/mol) 141.126
MDL Number MFCD00051661
InChI Key KLKQGSISBRVCTK-UHFFFAOYSA-N
Synonym methyl 5-amino-2-furoate, methyl5-amino-2-furoate, 5-aminofuran-2-carboxylic acid methyl ester, methyl 5-amino-2-furancarboxylate, 2-furancarboxylic acid, 5-amino-, methyl ester, methyl 5-amino-2-furate, acmc-1cq6f, methyl-2-amino-5-furoate, 2-amino-5-methoxycarbonylfuran, methyl5-aminofuran-2-carboxylate
PubChem CID 5072225
IUPAC Name methyl 5-aminofuran-2-carboxylate
SMILES COC(=O)C1=CC=C(O1)N

Specifications

Melting Point 131°C to 135°C
Quantity 1 g
Beilstein 1365799
Formula Weight 141.13
Percent Purity 98%
Chemical Name or Material Methyl 5-amino-2-furoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No

RUO – Research Use Only