missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-Acetylsalicylate 98.0+%, TCI America™

Catalog No. M165825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M165825G 25 g
M1658500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M165825G Supplier TCI America Supplier No. M165825G

Chemical Identifiers

CAS 16475-90-4
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
InChI Key XLSMGNNWSRNTIQ-UHFFFAOYSA-N
PubChem CID 85444
IUPAC Name methyl 5-acetyl-2-hydroxybenzoate
SMILES CC(=O)C1=CC(=C(C=C1)O)C(=O)OC

Specifications

Melting Point 60°C
Boiling Point 167°C
Quantity 25 g
Synonym methyl 5-acetylsalicylate, 5-acetylsalicylic acid methyl ester, 5-acetyl-2-hydroxybenzoic acid methyl ester, methyl 2-hydroxy-5-acetylbenzoate, benzoic acid, 5-acetyl-2-hydroxy-, methyl ester, 5-acetyl methyl salicylate, methyl 5-acethylsalicylate, pubchem22504, methyl5-acetylsalicylate, methyl-5-acetylsalicylate
Formula Weight 194.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-Acetylsalicylate