missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-acetylsalicylate, 98%

Catalog No. p-7080586
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1021514 25 g
AAA1021522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1021514 Supplier Thermo Scientific Chemicals Supplier No. A1021514
Only null left

CAS: 16475-90-4 | C10H10O4 | 194.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Methyl 5-acetylsalicylate is used in food flavorings and preservatives. It is useful in the relief of headache and muscle and joint aches It is also effective in reducing fever, inflammation, and swelling and thus has been used for treatment of rheumatoid arthritis, rheumatic fever, and mild infection.

Solubility
Soluble in methanol.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in well sealed containers.

Chemical Identifiers

CAS 16475-90-4
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00040924
InChI Key XLSMGNNWSRNTIQ-UHFFFAOYSA-N
Synonym methyl 5-acetylsalicylate, 5-acetylsalicylic acid methyl ester, 5-acetyl-2-hydroxybenzoic acid methyl ester, methyl 2-hydroxy-5-acetylbenzoate, benzoic acid, 5-acetyl-2-hydroxy-, methyl ester, 5-acetyl methyl salicylate, methyl 5-acethylsalicylate, pubchem22504, methyl5-acetylsalicylate, methyl-5-acetylsalicylate
PubChem CID 85444
IUPAC Name methyl 5-acetyl-2-hydroxybenzoate
SMILES CC(=O)C1=CC(=C(C=C1)O)C(=O)OC

Specifications

Melting Point 58°C to 62°C
Quantity 25 g
Beilstein 2643824
Solubility Information Soluble in methanol.
Formula Weight 194.19
Percent Purity 98%
Chemical Name or Material Methyl 5-acetylsalicylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 240-532-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only