missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-Acetamido-7,8, 9-tri-O-acetyl-5-N,4-O-carbonyl-3,5-dideoxy-2-S-phenyl-2-thio-D-glycero-β 96.0+%, TCI America™

Catalog No. p-7133159
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2319200MG 200 mg
M23191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2319200MG Supplier TCI America Supplier No. M2319200MG
Only null left

Methyl 5-Acetamido-7,8,9-tri-O-acetyl-5-N,4-O-carbonyl-3,5-dideoxy-2-S-phenyl-2-thio-D-glycero-beta-D-galacto-2-nonulopyranosylonate

Chemical Identifiers

CAS 934591-76-1
Molecular Formula C25H29NO12S
Molecular Weight (g/mol) 567.56
MDL Number MFCD22581442,MFCD22581442
InChI Key CVMYPHWPKCZUMN-UHFFFAOYNA-N
Synonym Methyl (Phenyl 5-Acetamido-7,8,9-tri-O-acetyl-5-N,4-O-carbonyl-3,5-dideoxy-2-thio-D-glycero-beta-D-galacto-2-nonulopyranosid)onate
PubChem CID 133554225
IUPAC Name methyl 3-acetyl-2-oxo-6-(phenylsulfanyl)-4-[1,2,3-tris(acetyloxy)propyl]-hexahydro-2H-pyrano[3,4-d][1,3]oxazole-6-carboxylate
SMILES COC(=O)C1(CC2OC(=O)N(C2C(O1)C(OC(C)=O)C(COC(C)=O)OC(C)=O)C(C)=O)SC1=CC=CC=C1

Specifications

Melting Point 152°C
Color White
Quantity 200 mg
Formula Weight 567.56
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 5-Acetamido-7,8,9-tri-O-acetyl-5-N,4-O-carbonyl-3,5-dideoxy-2-S-phenyl-2-thio-D-glycero-beta-
TSCA No
Recommended Storage Refrigerator